Learn More
asecos™ Stainless Steel Safety Canister
Product Code. 17806506 Shop All asecos Products
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
Height (Metric):
169.00 mm
241.00 mm
271.00 mm
280.00 mm
470.00 mm
Unit Size:
Each
Product Code. Capacity (Metric) Height (Metric) unitSize
17806506 10 L 241.00 mm Each
17826476 5 L 169.00 mm Each
17876536 10 L 280.00 mm Each
17816406 20 L 271.00 mm Each
17846486 20 L 470.00 mm Each
5 options
Product Code. 17806506
Brand: asecos™ 40091
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
  • Incl. multilingual safety sticker for content labelling, with filling tap made of stainless steel (1.4301)
  • Incl. ventilation, NBR seals and aluminium screw cap integrated flame arrester and plastic protective ring
Specifications
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Color Gray
Material Stainless Steel
Width (English) 9.61 in.
Width (Metric) 244.00 mm
Height (English) 9.49 in.
Height (Metric) 241.00 mm
Weight (English) 5.73 lb.
Weight (Metric) 2.6 kg