Learn More
OHAUS™ Stainless Steel Rod
Product Code. p-7158951 Shop All Ohaus Products
Click to view available options
Length (Metric):
40 mm
50 mm
150 mm
300 mm
460 mm
610 mm
910 mm
1220 mm
1520 mm
1830 mm
2440 mm
Unit Size:
Each
Product Code. 15772339
Brand: OHAUS™ 30392293
Only null left
Add to basket
Add to basket
OHAUS™ Stainless Steel Rod provides secure support when used with an OHAUS™ Lab Frame.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Dependable connectors secure rods and clamps to meet your requirements and five unique holders in a variety of sizes and materials accommodate different rods.
TRUSTED_SUSTAINABILITY
Specifications
Type Lab Frame Rod
Material Stainless Steel
Description Clamp, Rod, Steel 4 cm, CLR-RODS004
Diameter (Metric) 13 mm
Length (Metric) 40 mm