Learn More
Bochem™ Stainless Steel Rod
Product Code. 10070911
Change view
Click to view available options
Length (Metric):
1250 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10070911 1250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10070911 Supplier Bochem™ Supplier No. 5174
Estimated Shipment: 28-01-2027
Bochem™ Stainless Steel Rod is constructed of Stainless Steel and for use with stand bases.
  • Stainless Steel
Specifications
Material Stainless Steel (18/10)
Length (Metric) 1250 mm
Diameter (Metric) Outer 13 mm