Learn More
Schneider Gerd™ Stainless Steel Right Analysis Spoon Spatula
Description
- Spoons collect and transport dry substances or liquids
- Spatulas are ideal for working with powders, stirring and mixing
- Perfect for miniscule amounts of chemicals
- Two end shapes, offer increased versatility
- Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Specifications
| Quantity | 1 Ea. |
| Product Type | Right Analysis Spoon Spatula |
| Blade Material | Steel |
| Material | Stainless Steel |
| Length (Metric) | 180 mm |