Learn More
Grant Instruments™ Stainless Steel Perforated Base Tray
Description
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
- For use with SBB Aqua Plus and LSB Series Aqua Pro Series Unstirred Water Bath
- Choose from a vartiety of sizes.
Specifications
Specifications
| For Use With (Equipment) | DMU5, SAP2/5, JBN5 |
| Type | SBT6 Base Tray |