Learn More
Agencinox™ Stainless Steel Rectangular Tray
Product Code. p-8008861
Click to view available options
Dimensions (L x W x H):
205 x 130 x 20 mm
220 x 145 x 28 mm
245 x 175 x 30 mm
265 x 185 x 30 mm
285 x 185 x 30 mm
300 x 210 x 40 mm
355 x 255 x 40 mm
425 x 280 x 50 mm
510 x 340 x 50 mm
Unit Size:
Each
Product Code. 11751933
Brand: Agencinox™ 1220
Only null left
Add to basket
Add to basket
Use these trays for a variety of applications. Agencinox™ Stainless Steel Rectangular Trays are ideal for use in medical environments.
  • Rectangular tray
  • Stainless steel construction
  • Available in a variety of sizes
Specifications
Dimensions (L x W) 205 x 130 mm
Dimensions (L x W x H) 205 x 130 x 20 mm
Height (Metric) 20 mm
Length (Metric) 205 mm
Material Stainless Steel
Type Tray
Width (Metric) 130 mm
For Use With Instruments
Shape Rectangular