Learn More
Agencinox™ Stainless Steel Rectangular Tray
Product Code. 11771933
Change view
Click to view available options
Dimensions (L x W x H):
205 x 130 x 20 mm
220 x 145 x 28 mm
245 x 175 x 30 mm
265 x 185 x 30 mm
285 x 185 x 30 mm
300 x 210 x 40 mm
355 x 255 x 40 mm
425 x 280 x 50 mm
510 x 340 x 50 mm
Unit Size:
Each
Product Code. Dimensions (L x W x H) unitSize
11771933 245 x 175 x 30 mm Each
11751933 205 x 130 x 20 mm Each
11701943 300 x 210 x 40 mm Each
11781933 265 x 185 x 30 mm Each
11791933 285 x 185 x 30 mm Each
11711943 355 x 255 x 40 mm Each
11771943 425 x 280 x 50 mm Each
11781943 510 x 340 x 50 mm Each
11761933 220 x 145 x 28 mm Each
9 options
Product Code. 11771933
Brand: Agencinox™ 1224
Only null left
Add to basket
Add to basket
Use these trays for a variety of applications. Agencinox™ Stainless Steel Rectangular Trays are ideal for use in medical environments.
  • Rectangular tray
  • Stainless steel construction
  • Available in a variety of sizes
Specifications
Dimensions (L x W) 245 x 175 mm
Dimensions (L x W x H) 245 x 175 x 30 mm
Height (Metric) 30 mm
Length (Metric) 245 mm
Material Stainless Steel
Type Tray
Width (Metric) 175 mm
For Use With Instruments
Shape Rectangular