Learn More
Agencinox™ Stainless Steel Rectangular Tray
Description
- Rectangular tray
- Stainless steel construction
- Available in a variety of sizes
Specifications
Specifications
| Dimensions (L x W) | 205 x 130 mm |
| Dimensions (L x W x H) | 205 x 130 x 20 mm |
| Height (Metric) | 20 mm |
| Length (Metric) | 205 mm |
| Material | Stainless Steel |
| Type | Tray |
| Width (Metric) | 130 mm |
| For Use With | Instruments |
| Shape | Rectangular |