Learn More
Endecotts™ Stainless Steel Receiver
Click to view available options
Diameter (Metric):
200 mm
230 mm
450 mm
Unit Size:
Each
Description
Full height receivers for use with test sieves. Endecotts™ Stainless Steel Receivers are widely used with test sieves for all types of laboratory sampling and particle size analysis.
- Stainless steel construction
- For use with Stainless Steel Woven Wire Mesh Test Sieves
Specifications
Specifications
| Product Type | Receiver |
| Material | Stainless Steel |
| Diameter (Metric) | 200 mm |
| For Use With (Application) | For sieves |
| For Use With (Equipment) | Stainless Steel Woven Wire Mesh Test Sieve |
| Packaging Quantity | 1 Ea. |