Learn More
RETSCH Stainless Steel Push-fit Feed Chutes
Brand: RETSCH 03.729.0055
11651.75 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.25000kg
Specifications
Push-fit feed Chute | |
For ZM 200 | |
Stainless Steel |
270 x 40 mm | |
270 mm | |
40 mm |