Learn More
RETSCH Stainless Steel Push-fit Feed Chutes
Brand: RETSCH 03.729.0056
4845.00 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.20000kg
Specifications
Push-fit Feed Chute | |
For ZM 200 | |
Stainless Steel |
250 x 75/40 mm | |
250 mm | |
75/40 mm |