Specifications
| Product Type | Push-fit feed Chute |
| Dimensions (L x W) | 250 x 40 mm |
| For Use With (Equipment) | For ZM 200 |
| Length (Metric) | 250 mm |
| Material | Stainless Steel |
| Width (Metric) | 40 mm |