Learn More
RETSCH Stainless Steel Push-fit Feed Chutes
Brand: RETSCH 03.729.0036
4389.00 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.25000kg
Specifications
Push-fit Feed Chute | |
For Holder 15/40 DR 100 | |
Stainless Steel |
210 x 40 mm | |
210 mm | |
40 mm |