Learn More
Bochem™ Stainless Steel/PTFE Coated Standard Spoon Spatulas
Product Code. 10515801
Click to view available options
Length (Metric):
210 mm
230 mm
Unit Size:
Each
Product Code. 10515801
Brand: Bochem™ 3721
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Quantity 1 Ea.
Product Type Spoon Spatula
Diameter (Metric) Outer 4mm
Material Stainless Steel/PTFE Coated
Length (Metric) 210 mm