Learn More
Cole-Parmer™ Stainless Steel/Platinum RTD Immersion Probe
Product Code. 15267847 Shop All Cole-Parmer Products
Change view
Click to view available options
Type:
Probe
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15267847 Probe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15267847 Supplier Cole-Parmer™ Supplier No. 0464615
  • ONLINE EXCLUSIVE PRICE
    2010.00 NOK
    1821.78 NOK / Pack of 1
    Save 188.22 NOK
    9% Off
Estimated Shipment: 15-02-2027
For Use With: EchoTherm digital hot plates and stirring hot plates
Specifications
For Use With (Equipment) EchoTherm digital hot plates and stirring hot plates
Length (English) 6.5 in.
Type Probe