Learn More
Nickel Electro™ Stainless Steel Perforated Tray
Product Code. 11419549 Shop All Nickel-Electro Products
Change view
Click to view available options
No. of Positions:
12
18
2
2 x 1000 mL, 6 x 500 mL, 15 x 100 mL, 35 x 25 mL
32
39
5
8
Product Type:
Perforated Tray
Universal Spring Tray
Unit Size:
Each
Product Code. No. of Positions Product Type unitSize
11419549 2 x 1000 mL, 6 x 500 mL, 15 x 100 mL, 35 x 25 mL Universal Spring Tray Each
12798968 39 Perforated Tray Each
11736597 32 Perforated Tray Each
12708978 18 Perforated Tray Each
12718978 12 Perforated Tray Each
12728978 8 Perforated Tray Each
12738978 5 Perforated Tray Each
12748978 2 Perforated Tray Each
8 options
Product Code. 11419549
Brand: Nickel Electro™ SFT9000
Only null left
Add to basket
Add to basket
Nickel Electro™ Stainless Steel Perforated Tray is to be used with flask clips and can be used in a variety of baths.
  • Supplied ready to assemble
  • Stainless steel construction
  • Suits glass/polypropylene Erlenmeyer flasks
  • Tray simply drops onto shaker or rocking table platform and locates snugly
Specifications
Product Type Universal Spring Tray
Material Stainless Steel
Description Universal spring tray
For Use With (Equipment) NE5-28 Shaker Baths
No. of Positions 2 x 1000 mL, 6 x 500 mL, 15 x 100 mL, 35 x 25 mL