Learn More
De Buyer™ Stainless Steel Pastry Bowls
Product Code. 11990625
Change view
Click to view available options
Volume (Metric):
1 L
3.5 L
5.5 L
8 L
11.4 L
16.8 L
Height (Metric):
90 mm
100 mm
112 mm
125 mm
140 mm
150 mm
200 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Height (Metric) unitSize
11990625 1 L 100 mm Each
11980625 1 L 90 mm Each
11910605 3.5 L 112 mm Each
11920635 5.5 L 125 mm Each
11940635 8 L 140 mm Each
11950635 11.4 L 150 mm Each
11940645 16.8 L 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11990625 Supplier De Buyer™ Supplier No. 3250.20
  • ONLINE EXCLUSIVE PRICE
    253.05 NOK
    193.78 NOK / Pack of 1
    Save 59.27 NOK
    23% Off
 Restricted Item
Estimated Shipment: 07-09-2026
Specifications
Product Type Pastry Bowl
Material Stainless Steel
For Use With (Application) For making mixtures, mixing, Whisking, heating by bain-marie, for storing a preparation or ingredients
For Use With (Equipment) Fridge, deep-freezer, blast-chiller and oven
Height (Metric) 100 mm
Quantity Each
Shape Flat Bottom
Sterility Sterile
Volume (Metric) 1 L