Learn More
Cole-Parmer™ Essentials Stainless Steel Pail
Product Code. p-8004973 Shop All Cole-Parmer Products
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
11711836 Each
11794939 Each
11721836 Each
3 options
Product Code. 11711836
Brand: Cole-Parmer™ Essentials 0726022
Only null left
Add to basket
Add to basket
This Pail is constructed of heavy gauge, 304 stainless steel and features a flat pouring rim.
  • Minimize dripping with flat pouring rim
Specifications
Product Type Pail
Material Stainless Steel
Capacity (English) 3.2 gal.
Diameter (English) Outer 10 in.
Height (English) 12 in.
Height (English) With Closure 12 in.
Height (Metric) With Closure 304.8 mm
Quantity 1 Ea.