Learn More
Cole-Parmer™ Essentials Stainless Steel Pail
Product Code. 11794939 Shop All Cole-Parmer Products
Change view
Click to view available options
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
11794939 Each
11711836 Each
11721836 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11794939 Supplier Cole-Parmer™ Essentials Supplier No. 0727002
  • ONLINE EXCLUSIVE PRICE
    3540.00 NOK
    3422.74 NOK / Pack of 1
    Save 117.26 NOK
    3% Off
Estimated Shipment: 02-02-2027
This Pail is constructed of heavy gauge, 304 stainless steel and features a flat pouring rim.
  • Minimize dripping with flat pouring rim
Specifications
Capacity (English) 4 gal.
Diameter (English) Outer 10.1 in.
Height (English) 12 in.
Height (English) With Closure 12 in.