Learn More
Sartorius™ Stainless Steel OIML Class F2 Calibration Weight Without DAkkS Calibration Certificate
Product Code. 10042071 Shop All Sartorius Products
Change view
Click to view available options
Mass:
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
Each
Product Code. Mass unitSize
10042071 2 kg Each
10591541 100 g Each
10422491 200 g Each
10215701 10 kg Each
10432491 1 kg Each
10238510 500 g Each
10012121 5 kg Each
7 options
Product Code. 10042071
Brand: Sartorius™ YCW624800
Only null left
Add to basket
Add to basket
Highly polished steel calibration weights from 100g to 10kg
To OIML Class E2, F1 and F2 specification which includes DKD (Deutscher Kalibrierdienst accredited by the PTB) certificate (UKAS Equivalent).
TRUSTED_SUSTAINABILITY
Spezifikation
Case Material Plastic
Mass 2 kg