Learn More
- Home
- Products
- Balances and Scales
- Calibration Weights
- Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight Without DAkkS Calibration Certificate
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight Without DAkkS Calibration Certificate
Product Code. 10693922 Shop All Sartorius Products
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
50 kg
Unit Size:
Each
| Product Code. | Mass | unitSize |
|---|---|---|
| 10693922 | 200 mg | Each |
| 10173741 | 100 g | Each |
| 10052211 | 10 g | Each |
| 10642202 | 10 kg | Each |
| 10616274 | 10 mg | Each |
| 10656363 | 1 g | Each |
| 10397201 | 1 kg | Each |
| 10042131 | 1 mg | Each |
| 10032071 | 200 g | Each |
| 10387441 | 20 g | Each |
| 10745892 | 20 kg | Each |
| 10564181 | 2 kg | Each |
| 10612772 | 2 g | Each |
| 10275791 | 20 mg | Each |
| 10554371 | 2 mg | Each |
| 10490842 | 500 g | Each |
| 10327591 | 50 g | Each |
| 10706852 | 500 mg | Each |
| 10174411 | 50 kg | Each |
| 10585311 | 50 mg | Each |
| 10595501 | 5 g | Each |
| 10718772 | 5 kg | Each |
| 10662202 | 5 mg | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Stainless Steel OIML Class E2 Reference Line Weight Without DAkkS Calibration Certificate
Specifications
Specifications
| Case Material | Wood |
| Mass | 200 mg |