Learn More
- Home
- Products
- Balances and Scales
- Calibration Weights
- Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight Without DAkkS Calibration Certificate
Sartorius™ Stainless Steel OIML Class E2 Reference Line Weight Without DAkkS Calibration Certificate
Product Code. 10327591 Shop All Sartorius Products
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
50 kg
Unit Size:
Each
23 product options available for selection
| Product Code. | Mass | unitSize |
|---|---|---|
| 10327591 | 50 g | Each |
| 10173741 | 100 g | Each |
| 10052211 | 10 g | Each |
| 10642202 | 10 kg | Each |
| 10616274 | 10 mg | Each |
| 10656363 | 1 g | Each |
| 10397201 | 1 kg | Each |
| 10042131 | 1 mg | Each |
| 10032071 | 200 g | Each |
| 10693922 | 200 mg | Each |
| 10387441 | 20 g | Each |
| 10745892 | 20 kg | Each |
| 10564181 | 2 kg | Each |
| 10612772 | 2 g | Each |
| 10275791 | 20 mg | Each |
| 10554371 | 2 mg | Each |
| 10490842 | 500 g | Each |
| 10706852 | 500 mg | Each |
| 10174411 | 50 kg | Each |
| 10585311 | 50 mg | Each |
| 10595501 | 5 g | Each |
| 10718772 | 5 kg | Each |
| 10662202 | 5 mg | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Stainless Steel OIML Class E2 Reference Line Weight Without DAkkS Calibration Certificate
Specifications
Specifications
| Case Material | Wood |
| Mass | 50 g |