Learn More
Sartorius™ Stainless Steel OIML Class E2 Calibration Weight Without DAkkS Calibration Certificate
Product Code. 10298080 Shop All Sartorius Products
Click to view available options
Mass:
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
Unit Size:
Each
Product Code. 10298080
Brand: Sartorius™ YCW452800
Only null left
Add to basket
Add to basket
Highly polished steel calibration weights from 1g to 2kg
To OIML Class E2, F1 and F2 specification.
TRUSTED_SUSTAINABILITY
Specifications
Case Material Plastic
Mass 50 g