Learn More
Sartorius™ Stainless Steel OIML Class E2 Calibration Weight Without DAkkS Calibration Certificate
Product Code. 10084140 Shop All Sartorius Products
Change view
Click to view available options
Mass:
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
Unit Size:
Each
Product Code. Mass unitSize
10084140 5 kg Each
10309730 100 g Each
10309540 10 g Each
10627681 20 g Each
10399530 2 g Each
10379820 2 kg Each
10238180 1 kg Each
10688251 200 g Each
10591911 1 g Each
10166270 500 g Each
10298080 50 g Each
10106180 5 g Each
12 options
Product Code. 10084140
Brand: Sartorius™ YCW652800
Only null left
Add to basket
Add to basket
Highly polished steel calibration weights from 1g to 2kg
To OIML Class E2, F1 and F2 specification.
TRUSTED_SUSTAINABILITY
Specifications
Case Material Plastic
Mass 5 kg