Learn More
Sampling Systems™ Stainless Steel Multilevel Slot Sampler
Product Code. p-7138838 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
300 mm
600 mm
1000 mm
1500 mm
2000 mm
Capacity (Metric):
45 mL
110 mL
190 mL
300 mL
370 mL
750 mL
1000 mL
1300 mL
Diameter (Metric):
25 mm
40 mm
Includes:
3 Slots
3 Slots, 1 Bottle, 1 Adapter
4 Slots
4 Slots, 1 Bottle, 1 Adapter
5 Slots
5 Slots, 1 Bottle, 1 Adapter
Unit Size:
Each
Product Code. Length (Metric) Capacity (Metric) Diameter (Metric) Includes unitSize
15252760 300 mm 45 mL 25 mm 3 Slots Each
15879221 1000 mm 750 mL 40 mm 3 Slots Each
15809221 600 mm 110 mL 25 mm 3 Slots Each
15819221 600 mm 190 mL 25 mm 4 Slots, 1 Bottle, 1 Adapter Each
15829221 1500 mm 300 mL 25 mm 5 Slots, 1 Bottle, 1 Adapter Each
15839221 600 mm 110 mL 25 mm 3 Slots, 1 Bottle, 1 Adapter Each
15879211 1000 mm 190 mL 25 mm 4 Slots Each
15889211 1500 mm 300 mL 25 mm 5 Slots Each
15889221 1500 mm 1000 mL 40 mm 4 Slots Each
15899211 2000 mm 370 mL 25 mm 5 Slots Each
15899221 2000 mm 1300 mL 40 mm 4 Slots Each
11 options
Product Code. 15252760
Brand: Sampling Systems™ 1210A300
Only null left
Add to basket
Add to basket
Constructed of robust stainless steel, making it suitable for sampling free flowing powders, granules and even slightly cohesive powders. Sampling Systems™ Stainless Steel Multilevel Slot Sampler is for taking a large volume, cross sectional sample from a container.
  • Made from high quality 316L stainless steel
  • Cross section (Composite sample)
  • Suitable for taking large volume samples
Specifications
Product Type Slot Sampler
Material Stainless Steel (316L)
Includes 3 Slots
Capacity (Metric) 45 mL
Diameter (Metric) 25 mm
Length (Metric) 300 mm