Learn More
Sampling Systems™ Stainless Steel Mini-Drums without Toggle Clamps
Product Code. 15899661 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
1 L
2 L
3 L
5 L
6 L
8 L
10 L
12.5 L
15 L
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15899661 1 L Each
15839671 2 L Each
15849671 3 L Each
15859671 5 L Each
15869671 6 L Each
15879671 8 L Each
15809671 10 L Each
15819671 12.5 L Each
15829671 15 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15899661 Supplier Sampling Systems™ Supplier No. A4341
  • ONLINE EXCLUSIVE PRICE
    3020.00 NOK
    2903.23 NOK / Pack of 1
    Save 116.77 NOK
    3% Off
Estimated Shipment: 23-12-2026
These Mini-Drums without Toggle Clamps are excellent multi-purpose containers for a wide variety of applications in the laboratory.
  • Manufactured from durable 304 grade stainless steel
  • Crevice-free interior
  • Neat recessed fitting lid
  • Complete with a swing carrying handle
  • Flat bottom with chime ring to give added protection
  • Available in a wide range of capacities to fit your specific needs
Specifications
Closure Included Yes
Product Type Mini Drum
Closure Type Recessed Lid
Capacity (Metric) 1 L
For Use With (Application) Multi-purpose Storage
Material Stainless Steel
Includes Swing Handle
Height (Metric) 155 mm
Diameter (Metric) Outer 100 mm