Learn More
Schneider Gerd™ Stainless Steel Micro-Spoon Spatula
Product Code. p-8000771
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Product Code. 10798112
Brand: Schneider Gerd™ 252
Only null left
Add to basket
Add to basket
Use this micro spatula and spoon for a variety of applications. Schneider Gerd™ Stainless Steel Micro-Spoon Spatulas are ideal for use with a wide range of compounds.
  • Micro-spoons collect and transport dry substances or liquids
  • Spatulas are ideal for working with powders, stirring and mixing
  • Perfect for working with miniscule amounts of chemicals
  • Two different end shapes, offer increased versatility
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Quantity 1 Ea.
Shape Spoon
Product Type Micro Spoon Spatula
Width (Metric) 6 mm
Material Stainless Steel
Length (Metric) 150 mm