Learn More
Hamilton™ Custom Kel-F Hub Needles
Description
- Available in various gauges
- Provided in packs of 6
- Custom point style
- Autoclavable
- Gas sterilizable
- For Kel-F hubs
- Custom length
- Stainless steel Luer tip
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Needle Gauge | 17 G |
| Diameter (Metric) Inner | 1.07 mm |
| Diameter (Metric) Outer | 1.47 mm |
| Product Type | Luer Lock Kel-F Hub Needle |
| Quantity | 6/Pk. |
| Needle Point Style | 2, 3, or 4 |
| For Use With (Equipment) | LT and TLL Syringes and Connectors |
| Connection Type | Standard Tapered |