Learn More
Cole-Parmer™ Stainless Steel Luer Fittings
Click to view available options
Includes:
Female Luer x 1/4-28 UNF Thread
Female Luer x Male Luer Lock
Male LL x 1/4-28 UNF Thread
Male LL x 1/4in. Hose Barb
Unit Size:
Each
Description
Exceptionally durable Luer fittings suitable for harsh chemical environments. Cole-Parmer™ Stainless Steel Luer Fittings are easy to disassemble for cleaning.
- Exceptional durability
- Easy to disassemble for cleaning
- Ideal for harsh chemical environments
- Autoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| For Use With (Application) | Harsh chemical environments |
| Packaging Quantity | 1 Pack |
| Includes | Male LL x 1/4-28 UNF Thread |
| Product Type | Luer Fitting |