Learn More
Grant Instruments™ Stainless Steel Lid with Ring Sets
Product Code. 11466427
Click to view available options
For Use With (Equipment):
JBA12, JBN12, SAP12
JBA18, JBN18, SAP18
JBA5, JBN5, SAP5
SAP34
Unit Size:
Each
Product Code. 11466427
Brand: Grant Instruments™ LF14
Only null left
Add to basket
Add to basket
Accommodate tall vessels whilst reducing evaporation. Grant Instruments Stainless Steel Lid with Ring Sets are flat with a range of variable hole diameters.
  • Stainless steel construction
  • Flat lids contain 2, 4 or 6 ring sets.
  • Variable hole diameter
  • Accommodates taller vessels
TRUSTED_SUSTAINABILITY
Specifications
Description 4-Ring Sets
For Use With (Equipment) JBA12, JBN12, SAP12
Material Stainless Steel