Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Description
- Available in various gauges
- Provided in packs of 6
- Custom point style and lengths
- Autoclavable
- For large hubs
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Needle Gauge | 23s G |
| Diameter (Metric) Inner | 0.12 mm |
| Diameter (Metric) Outer | 0.64 mm |
| Product Type | Removable Custom Needle |
| Needle Point Style | 5 or AS |
| For Use With (Equipment) | Removable Syringes (250μL and larger) |