Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Click to view available options
Needle Gauge:
22 G
22s G
23 G
23s G
25s G
26 G
26s G
Needle Point Style:
2, 3, or 4
5 or AS
Unit Size:
Pack of 6
Description
- Available in various gauges
- Provided in packs of 6
- Custom point style and lengths
- Autoclavable
- For large hubs
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Needle Gauge | 23 G |
| Diameter (Metric) Inner | 0.34 mm |
| Diameter (Metric) Outer | 0.64 mm |
| Product Type | Removable Custom Needle |
| Needle Point Style | 2, 3, or 4 |
| For Use With (Equipment) | Removable Syringes (250μL and larger) |