Learn More
Sampling Systems™ Stainless Steel Ladles
Product Code. p-7146966 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
50 mL
120 mL
150 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12987955 50 mL Each
12967955 120 mL Each
12977955 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12987955 Supplier Sampling Systems™ Supplier No. A32450
These ladles are made from hygienic stainless steel and are ideal for sampling liquid products.
  • Satin finish
  • Crevice-free construction so they can be easily cleaned between batches
  • Range of sizes available
Specifications
Product Type Ladle
Material Stainless Steel (AISI 304)
Capacity (Metric) 50 mL
For Use With (Application) Sampling liquid products
Length (Metric) 275 mm