Learn More
Sampling Systems™ Stainless Steel Ladles
Product Code. 12987955 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
50 mL
120 mL
150 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
12987955 50 mL Each
12967955 120 mL Each
12977955 150 mL Each
3 options
Product Code. 12987955
Brand: Sampling Systems™ A32450
Only null left
Add to basket
Add to basket
These ladles are made from hygienic stainless steel and are ideal for sampling liquid products.
  • Satin finish
  • Crevice-free construction so they can be easily cleaned between batches
  • Range of sizes available
Specifications
Product Type Ladle
Material Stainless Steel (AISI 304)
Capacity (Metric) 50 mL
For Use With (Application) Sampling liquid products
Length (Metric) 275 mm