Learn More
Bochem™ Stainless Steel Laboratory Scissors with Plastic Handle
Product Code. 10084900
Change view
Click to view available options
Length (Metric):
130 mm
150 mm
200 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10084900 150 mm Each
10288780 130 mm Each
10300921 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10084900 Supplier Bochem™ Supplier No. 4021
Estimated Shipment: 10-11-2026
Use the Bochem™ Stainless Steel Laboratory Scissors with Plastic Handle to handle all of your cutting and trimming needs in the laboratory.
  • Scissors are Type 2, with one sharp and one round cutting tip each
  • Stainless steel
  • Plastic handle
Specifications
Product Type Laboratory Scissors
Length (Metric) 150 mm
Material Stainless Steel
Tip Style Sharp/Round