Learn More
Bochem™ Stainless Steel Laboratory Pot
Description
Note:
Lid is not included; sold separately.
Specifications
Specifications
| Diameter (Metric) Inner | 160 mm |
| Height (Metric) | 128H |
| Capacity (Metric) | 2.5 L |
| Material | Stainless Steel (18/10) |
| Product Type | Laboratory Pot |