Learn More
Bochem™ Stainless Steel Laboratory Pot
Product Code. 10606721
Change view
Click to view available options
Capacity (Metric):
2.5 L
3.5 L
5 L
6.5 L
8.5 L
15.5 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10606721 3.5 L Each
10666331 2.5 L Each
10135840 5 L Each
10258320 6.5 L Each
10669852 8.5 L Each
10653072 15.5 L Each
6 options
Product Code. 10606721
Brand: Bochem™ 8361
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Note:
Lid is not included; sold separately.
Specifications
Diameter (Metric) Inner 180 mm
Height (Metric) 144H
Capacity (Metric) 3.5 L
Material Stainless Steel (18/10)
Product Type Laboratory Pot