Learn More
Nickel Electro™ Stainless Steel Beakers with Spout
Product Code. 11459569 Shop All Nickel-Electro Products
Change view
Click to view available options
Capacity (Metric):
200 mL
350 mL
550 mL
700 mL
1000 mL
1250 mL
2000 mL
2500 mL
5000 mL
Unit Size:
Each
Pack of 5
Product Code. Capacity (Metric) unitSize
11459569 2500 mL Each
11756567 200 mL Each
12737856 350 mL Each
12747856 550 mL Each
11766617 550 mL Pack of 5
11419569 700 mL Each
11429569 1000 mL Each
11439569 1250 mL Each
11449569 2000 mL Each
11469569 5000 mL Each
10 options
Product Code. 11459569
Brand: Nickel Electro™ 208
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Will not shatter or chip, this durable stainless steel laboratory beaker is suitabale for laboratory applications.
  • Durable stainless steel laboratory beaker
  • 304 spec stainless steel beakers 
  • Will not shatter or chip
  • Suitable for laboratory applications since 
  • All surfaces are non-reacting 
  • Offer zero absorption
Specifications
Diameter (Metric) Outer 150 mm
Capacity (Metric) 2500 mL
Material Stainless Steel
Height (Metric) 160 mm
Includes Spout
Type Beaker