Learn More
Nickel Electro™ Stainless Steel Beakers with Spout
Product Code. 11429569 Shop All Nickel-Electro Products
Change view
Click to view available options
Capacity (Metric):
200 mL
350 mL
550 mL
700 mL
1000 mL
1250 mL
2000 mL
2500 mL
5000 mL
Unit Size:
Each
Pack of 5
Product Code. Capacity (Metric) unitSize
11429569 1000 mL Each
11756567 200 mL Each
12737856 350 mL Each
12747856 550 mL Each
11766617 550 mL Pack of 5
11419569 700 mL Each
11439569 1250 mL Each
11449569 2000 mL Each
11459569 2500 mL Each
11469569 5000 mL Each
10 options
Product Code. 11429569
Brand: Nickel Electro™ 206
Only null left
Add to basket
Add to basket
Will not shatter or chip, this durable stainless steel laboratory beaker is suitabale for laboratory applications.
  • Durable stainless steel laboratory beaker
  • 304 spec stainless steel beakers 
  • Will not shatter or chip
  • Suitable for laboratory applications since 
  • All surfaces are non-reacting 
  • Offer zero absorption
Specifications
Diameter (Metric) Outer 110 mm
Capacity (Metric) 1000 mL
Material Stainless Steel
Height (Metric) 135 mm
Includes Spout
Type Beaker