Learn More
OHAUS™ Stainless Steel Lab Frame
Product Code. p-7158942 Shop All Ohaus Products
Change view
Click to view available options
Size:
305 x 1829 x 1219 mm (LxWxH)
457 x 1219 x 1219 mm (LxWxH)
457 x 1219 x 610 mm (LxWxH)
457 x 610 x 610 mm (LxWxH)
551 x 1829 x 1905 mm (LxWxH)
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15772369 457 x 610 x 610 mm (LxWxH) Each
15782369 457 x 1219 x 610 mm (LxWxH) Each
15792369 457 x 1219 x 1219 mm (LxWxH) Each
15702379 305 x 1829 x 1219 mm (LxWxH) Each
15722379 551 x 1829 x 1905 mm (LxWxH) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15772369 Supplier OHAUS™ Supplier No. 30392323
Only null left
Add to basket
Add to basket
OHAUS™ Stainless Steel Lab Frame provides secure support for holding glassware at elevated positions.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Dependable connectors secure rods and clamps to meet your requirements and five unique holders in a variety of sizes and materials accommodate different rods.
TRUSTED_SUSTAINABILITY
Specifications
Type Lab Frame Kit
Material Stainless Steel
Description Clamp, Lab Frame, Steel, CLR-FRAMESS
Height (Metric) 610.000 mm
Height (English) 24.00 in.
Length (English) 18.00 in.
Length (Metric) 457.00 mm
Size 457 x 610 x 610 mm (LxWxH)
Width (English) 24.00 in.
Width (Metric) 610.00 mm