Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Dependable connectors secure rods and clamps to meet your requirements and five unique holders in a variety of sizes and materials accommodate different rods.
Specifications
Specifications
| Type | Lab Frame Kit |
| Material | Stainless Steel |
| Description | Clamp, Lab Frame, Steel, CLR-FRAMESSL |
| Height (Metric) | 1,219.000 mm |
| Height (English) | 48.00 in. |
| Length (English) | 18.00 in. |
| Length (Metric) | 457.00 mm |
| Size | 457 x 1219 x 1219 mm (LxWxH) |
| Width (English) | 48.00 in. |
| Width (Metric) | 1,219.00 mm |