Learn More
Retsch™ Stainless Steel Knife
Product Code. 15383278 Shop All Retsch Products
Change view
Click to view available options
Product Type:
Metal Blade and Knife Cylinder
Serrated Blade
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
15383278 Serrated Blade Each
15860530 Metal Blade and Knife Cylinder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15383278 Supplier Retsch™ Supplier No. 02.446.0054
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
Retsch™ Stainless Steel Knife is designed for use with Knife Mill GRINDOMIX GM 300.
  • Constructed of stainless steel
Specifications
Product Type Serrated Blade
For Use With (Equipment) Knife Mill GRINDOMIX GM 300
Material Stainless Steel