Learn More
Nickel Electro™ Stainless Steel Jugs
Product Code. p-8013894 Shop All Nickel-Electro Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
3000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
12827595 500 mL Each
11439609 1000 mL Each
12837595 2000 mL Each
15311005 3000 mL Each
4 options
Product Code. 12827595
Brand: Nickel Electro™ JUGS0500H
Only null left
Add to basket
Add to basket
304 spec stainless steel jugs are suitable for pharmaceutical and laboratory applications.
  • 304 spec stainless steel seamless jugs
  • Suitable for pharmaceutical and laboratory applications
Specifications
Diameter (Metric) Outer 90 mm
Capacity (Metric) 500 mL
Material Stainless Steel
Finish Bright
Height (Metric) 135 mm
Type Jug
Graduated Yes