Learn More
Justrite™ Stainless Steel Joining Clips for Connecting EcoPolyBlend™ Accumulation Centers
Product Code. 11919395
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Pack of 2
Product Code. Material unitSize
11919395 Stainless Steel Pack of 2
1 options
Product Code. 11919395
Brand: Justrite™ 28926
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bring old equipment back into service with replacement parts. The Justrite™ Stainless Steel Joining Clips for Connecting EcoPolyBlend™ Accumulation Centers is an accessory to help facilitate the use of accumulation centers.
By joining centers together with these clips or the patented Sump-to-Sump Drain™ kit for a total 66 gal. sump capacity, you will meet EPA and UFC requirements.
For compliant storage using unites 4-drum and smaller, use these joining clips to reach needed capacity. Also available as a kit (19-822-446).
Specifications
Color Silver
Material Stainless Steel
Width (English) 2 in.
Height (English) 4 in.