Learn More
Bochem™ Stainless Steel Instruments Tray
Product Code. 10253981
Click to view available options
Dimensions (L x W x H):
160 x 100 x 30 mm
200 x 100 x 50 mm
220 x 150 x 50 mm
240 x 180 x 40 mm
270 x 210 x 40 mm
280 x 180 x 60 mm
300 x 200 x 15 mm
300 x 200 x 50 mm
360 x 300 x 40 mm
440 x 340 x 20 mm
500 x 200 x 80 mm
300 x 150 x 100 mm
Unit Size:
Each
Product Code. 10253981
Brand: Bochem™ 8716
Only null left
Specifications
Dimensions (L x W) 300 x 200 mm
Dimensions (L x W x H) 300 x 200 x 50 mm
Height (Metric) 50 mm
Length (Metric) 300 mm
Material Stainless Steel
Type Instrument Tray
Width (Metric) 200 mm
Includes Lid
Quantity 1 Each
Shape Rectangular