Learn More
Sampling Systems™ Stainless Steel Instrument Trays
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
215 mm
295 mm
395 mm
490 mm
Width (Metric):
160 mm
197 mm
275 mm
320 mm
Height (Metric):
11 mm
15 mm
18 mm
Unit Size:
Each
Description
- Range of sizes
- Crevice free design
- High quality polished finished
- 304 Stainless Steel
Specifications
Specifications
| Height (Metric) | 18 mm |
| Length (Metric) | 295 mm |
| Material | Stainless Steel (AISI 304) |
| Type | Tray |
| Width (Metric) | 197 mm |
| Finish | Polished |