Learn More
Sampling Systems™ Stainless Steel Instrument Trays
Product Code. 15879701 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
215 mm
295 mm
395 mm
490 mm
Width (Metric):
160 mm
197 mm
275 mm
320 mm
Height (Metric):
11 mm
15 mm
18 mm
Unit Size:
Each
Product Code. 15879701
Brand: Sampling Systems™ A674215
Only null left
Add to basket
Add to basket
Features a crevice free design that allows for easy cleaning. Sampling Systems™ Stainless Steel Instrument Trays are ideal for carrying tools and equipment.
  • Range of sizes
  • Crevice free design
  • High quality polished finished
  • 304 Stainless Steel
Specifications
Height (Metric) 18 mm
Length (Metric) 215 mm
Material Stainless Steel (AISI 304)
Type Tray
Width (Metric) 160 mm
Finish Polished