Learn More
Socorex™ Stainless Steel Injection Needle
Click to view available options
Diameter (Metric) Outer:
1 mm
1.2 mm
Unit Size:
Each
Pack of 12
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Needle Gauge | 19 G |
| Diameter (Metric) Outer | 1 mm |
| Length (English) Needle | 3/4 in. |
| Product Type | Injection Needle |