Description
- Stainless steel AISI 316L (1.4404) / AISI 316 Ti (1.4571)
- Sterilisable
- 20 cm boring depth
- The sharp sawing crown of the very robust hollow borer rapidly cuts into the sample
- A cylindrical sample (cross-sectional sample) of up to 20 cm can be cut out
Specifications
Specifications
| Product Type | Ice Borer |
| Material | Stainless Steel (V4A) |
| Autoclavable | Autoclavable |
| Description | I.D. 50 mm |
| For Use With (Application) | Ideal for Deep-Frozen and Semi-Solid Substances Products in The Food Industry |
| Diameter (Metric) Outer Top | 54 mm |
| Length (Metric) | 200 mm |