missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, High Nitrogen, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Stainless Steel, High Nitrogen, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Nitrogen/Molybdenum/Carbon/Manganese/Silicon/Nickel/Chromium |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Nitrogen 0.04 %/Molybdenum 0.11 %/Carbon 0.17 %/Manganese 0.44 %/Silicon 0.58 %/Nickel 10 %/Chromium 19.1 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction